Learn More
Eppendorf™ PCR Films and Foils
Provide your PCR plate with a tight seal, particularlty when when heat sealing material is used. Eppendorf™ PCR Films and Foils feature superior optical characteristics, making them the ideal sealing material for qPCR and other optical assays.
Brand: Eppendorf™ 0030127781
144.00 GBP valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.36000kg
Description
- Effective adhesive sealing prevents evaporation loss during PCR
- Removal from the plate with no residue
- Sample can be visually monitored through the transparent PCR film
- PCR Foil: Easily pierced
- PCR Foil: No sticking of the pipette tip (ideal for automated systems)
Specifications
Sterile | |
100 | |
PCR Films and Foils | |
Yes |
Autoclavable | |
Ideal for Automated Systems Like PCR, RT PCR | |
Self | |
121°C |