Learn More
Eppendorf™ PCR Foil
Use this foil for PCR and other optical assays. Eppendorf™ PCR Foils provide your PCR plate with a tight seal, particularly when heat-sealing material is used.
Brand: Eppendorf™ 0030127790
144.00 GBP valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.30000kg
Description
- Effective adhesive sealing prevents evaporation loss during PCR
- Removal from the plate with no residue
- Easily pierced
- No sticking of the pipette tip (ideal for automated systems)
Specifications
Sterile | |
Ideal for Automated Systems, PCR, RT PCR | |
PCR Films and Foils | |
Yes |
100 Pk. | |
Ideal for Automated Systems Like PCR, RT PCR | |
Self | |
121°C |