missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ PCR Nucleotide Mix
A premixed 10mM solution of dATP, dCTP, dGTP, and dTTP used for PCR
£60.85 - £303.00
Specifications
Format | Liquid |
---|---|
pH | 7.5 |
Content And Storage | -20°C |
Concentration | 10 mM |
Purity | >98% |
Description
- Functionally tested in PCR
- High purity: >98% triphosphate
- Supplied as 10mM solution in purified water (pH 7.5)
Specifications
Liquid | |
-20°C | |
>98% | |
PCR |
7.5 | |
10 mM | |
Colorless |