Learn More
Applied Biosystems™ PCR Product Pre-Sequencing Kit
The PCR Product Pre-Sequencing Kit uses a novel, enzymatic clean-up method to pre-treat PCR products prior to sequencing, without any subsequent purification or separation steps.
Brand:  Applied Biosystems™ 70997.1.KT
Product Code. 15849896