missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ PCR Product Pre-Sequencing Kit
Click to view available options
No. of Reactions:
100
2000
500 Reactions
Unit Size:
100 reactions
2000 reactions
500 reactions
Description
- Remove unincorporated primers and nucleotides in about 5 minutes
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery, regardless of amplicon length
- Minimize spin column or bead clean-up
Specifications
Specifications
| Content And Storage | Store at -5 to -30°C. |
| Description | PCR Product Pre-Sequencing Kit, 100 rxns, Recombinant, PCR Purification, Uses a Novel, Enzymatic Clean-up Method to Pre-treat PCR Products Prior to Sequencing, without Any Subsequent Purification or Separation Steps |
| For Use With (Application) | PCR Purification |
| No. of Reactions | 100 |
| Product Line | ExoSAP-IT |
| Protein Form | Recombinant |
| Quantity | 100 reactions |
| Research Category | Genetic Diseases |
| Shipping Condition | Dry Ice |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction