Learn More
Brady™ PCR Tube Label for BBP33/i3300/i5300 Printers
The B-492 PCR tube label contains 8 smaller, perforated labels in a single label strip that can be applied on 8 PCR tubes in a rack. It can easily be torn into separate labels thanks to the perforations.
Brand:  Brady™ 307018
Product Code. 30041450
  • £170.00 / Pack of 1500
Explore more special offers
Description
Description
For forations.
Specifications
Specifications
Permanent Acrylic
Matte
Polyester (B-492)
Labels
9.50 mm (W) x 6.35 mm (H)
9.50 mm
White
Roll
Thermal Transfer
Non-sterile
6.35 mm
9.50 mm