missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PCR Tube Strips
Extra-thin and even wall thicknesses allow for optimal heat transfer and short cycle times. Smooth shaft surfaces minimize enzyme and nucleic acid binding to the surface ensuring the reaction can run in full.
Brand: BRAND™ 781327
194.87 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polypropylene 8 and 12 tube strips
- A variety of cap styles is available
- Compatible with leading thermal cyclers with heated lids
- Provide tight seal in all PCR* cyclers
- Protect samples from cross contamination
Specifications
Clear | |
250 Pack | |
Separate Domed |
0.2 mL | |
Polypropylene | |
PCR Tube Strip |