missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PCR Tubes

Constructed of extra-thin wall thickness to provide optimal heat transfer and reduced cycle time. Fisherbrand™ PCR Tubes eliminate contamination and sample loss with high-quality raw materials.

Brand:  Fisherbrand™ FI3-010-50-0

Product Code. 10385572

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Reduced cycle time and optimal heat transfer - guaranteed by extra thin, uniform wall thickness of tube / available with flat or domed cap
  • Effortless and secure working - ergonomic cap design allows opening and closing with one hand
  • Made of materials that meets standards of purity
  • Highly resistant to extreme mechanical strain with excellent chemical resistance
  • Optical measurement and unsurpassed clarity
  • Maintain sample integrity - sterile version available (beta irradiation)
  • Sterile handling - PCR tubes are autoclavable
  • Centrifuge specifications:
    • up to 25.000 x g - fixed-angle rotor
    • up to 30.000 x g - swing out rotor
Specifications

Specifications

CE, IVD
0.2 mL
Centrifuge
Flat Cap
PCR Tube
Clear
1000 Pack
Polypropylene
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ PCR Tubes > Color: Clear; Volume: 0.2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.