missing translation for 'onlineSavingsMsg'
Learn More

PCR WaterAbnova™

Product Code. 16062565
Click to view available options
Quantity:
10 x 1.8 mL
25 x 1.8 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Manufactured and tested for use as a component in PCR reactions, Validated as well for use with highly sensitive PCR reaction including those with eubacterial primer sets.

Stored in convenient 2 mL tubes to avoid unnecessary melting and freezing of the whole water supply.

Spezifikation

Quantity 10 x 1.8 mL
Produktidentifikator
  • X10 PCR Water 1.8 mL
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'