missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-1 Monoclonal Antibody (J43), PE, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen PA535011
This item is not returnable.
View return policy
Description
PA5-35011 targets CD279/PD1 in FACS applications and shows reactivity with Mouse samples. The PA5-35011 immunogen is pD-1 cDNA transfectant.
Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "e;danger signals"e; such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases. Despite its predicted molecular weight, PD-1 often migrates at higher molecular weight in SDS-PAGE.Specifications
PD-1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Q02242 | |
Pdcd1 | |
PD-1 cDNA transfectant | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry | |
J43 | |
PE | |
Pdcd1 | |
CD279; EGK_05005; hPD1; hPD-1; hPD-l; hSLE1; Ly101; mPD-1; PD1; PD-1; Pdc1; Pdcd1; programmed cell death 1; programmed cell death 1 protein; programmed cell death protein 1; programmed cell death protein 1-like; programmed death 1; Protein PD1; protein PD-1; sCD279; SLEB2; soluble CD279; systemic lupus erythematosus susceptibility 2 | |
Armenian Hamster | |
Protein G | |
RUO | |
18566 | |
4° C, do not freeze | |
Liquid |