missing translation for 'onlineSavingsMsg'
Learn More

PD 168077 maleate, Tocris Bioscience™

Product Code. 18084692
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity, selective D4 agonist

A potent D4 dopamine receptor agonist (Ki = 8.7 nM) with > 400-fold selectivity over D2 and > 300-fold selectivity versus D3 subtypes respectively. Induces synaptic translocation of CaMK II to postsynaptic sites in cultured prefrontal cortical neurons. Centrally active in vivo.

Chemical Identifiers

CAS 630117-19-0
Molecular Formula C24H26N4O5
Molecular Weight (g/mol) 450.495
InChI Key NAEUGRPISCANHO-BTJKTKAUSA-N
Synonym pd 168077 maleate, pd 168,077 maleate, n-4-2-cyanophenyl piperazin-1-yl methyl-3-methylbenzamide maleate, pd 168,077 maleate salt, n-4-2-cyanophenyl piperazin-1-yl methyl-3-methylbenzamide; maleic acid, n-4-2-cyanophenyl-1-piperazinyl methyl-3-methylbenzamide maleate salt, pd168077 maleate, pd 168,077 maleate salt, powder, n-methyl-4-2-cyanophenyl piperazinyl-3-methylbenzamide maleate, n-4-2-cyanophenyl-1-piperazinyl methyl-3-methyl-benzamide maleate
PubChem CID 11957665
IUPAC Name (Z)-but-2-enedioic acid;N-[[4-(2-cyanophenyl)piperazin-1-yl]methyl]-3-methylbenzamide
SMILES CC1=CC=CC(=C1)C(=O)NCN2CCN(CC2)C3=CC=CC=C3C#N.C(=CC(=O)O)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 450.49
Percent Purity 98%
Chemical Name or Material N-[[4-(2-Cyanophenyl)-1-piperazinyl]methyl]-3-methylbenzamide maleate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'