missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PD 90780
Inhibits NGF binding to p75NTR
£198.00 - £755.00
Specifications
Purity | >98% |
---|---|
Product Type | PD 90780 |
Molecular Weight (g/mol) | 362.34 |
Inhibitors | PD 90780 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18756158
|
Tocris Bioscience™
5101/50 |
50 mg |
£755.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18737088
|
Tocris Bioscience™
5101/10 |
10 mg |
£198.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A nonpeptide inhibitor of NGF binding to p75NTR (IC50 ∽ 1 μM in CHO cells); binds NGF, not p75NTR. Affinity maybe altered by the presence of TrkA receptors. Neuroprotective.Specifications
>98% | |
PD 90780 | |
362.34 | |
PD 90780 |
For Research Use Only