Learn More
BRAND™ PD-Tip™ II Precision Dispenser Tips for Repeating Pipettes
BRAND™ PD-Tip™ II fits most instruments using standard repeating tips.
£29.65 - £112.00
Specifications
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Not Autoclavable
Model PD-Tip II
View More Specs
Products 10
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
16368056
View Documents Promotion Details
BRAND™
705700
0.1 mL
£112.00
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16323166
View Documents Promotion Details
BRAND™
705702
0.5 mL
£96.90
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16333166
View Documents Promotion Details
BRAND™
705704
1 mL
£96.90
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16343166
View Documents Promotion Details
BRAND™
705706
1.25 mL
£96.90
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16353166
View Documents Promotion Details
BRAND™
705708
2.5 mL
£96.90
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16363166
View Documents Promotion Details
BRAND™
705710
5 mL
£96.90
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16373166
View Documents Promotion Details
BRAND™
705712
10 mL
£96.90
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16309635
View Documents Promotion Details
BRAND™
705714
12.5 mL
£96.90
Pack of 100
In Stock
to see stock.
Edge
Add to basket
16383166
View accessory products View Documents Promotion Details
BRAND™
705716
25 mL
£54.75
Pack of 50
In Stock
to see stock.
Edge
Add to basket
16393166
View accessory products View Documents Promotion Details
BRAND™
705718
50 mL
£29.65
Pack of 25
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Automatic size recognition by BRAND™ HandyStep™ Touch, HandyStep™ Touch S and HandyStep™ Electronic Pipettes
  • Adapter included for 25 mL and 50 mL sizes
  • Adapters are autoclavable at 121°C for 20 minutes
  • PD-Tips II are not autoclavable
These tips are suitable for the following dispensers:
  • BRAND Handystep™, BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch, HandyStep™ touch S
  • Eppendorf Multipette™ 4780
  • Gilson Repetman™EDOS™ 5221
  • Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
Specifications
Specifications
Non-filtered
Natural
PD-Tip II
Virgin raw materials
Single-use
Yes
Polypropylene, Polyethylene
Non-sterile
Not Autoclavable
Certificate of Performance
Positive Displacement
Universal Fit Repeater Pipettor
PD-Tip II
Printed
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BRAND™ PD-Tip™ II Precision Dispenser Tips for Repeating Pipettes