missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDF Monoclonal Antibody (OTI3F3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA525562
This item is not returnable.
View return policy
Description
Removes the formyl group from the N-terminal Met of newly synthesized proteins.Specifications
Monoclonal | |
0.78 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9HBH1 | |
Full length human recombinant protein of PDF produced in E.coli | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI3F3 | |
Unconjugated | |
2610019N19Rik; PDF; PDF1A; peptide deformylase (mitochondrial); peptide deformylase, mitochondrial; peptide deformylase-like protein; peptide deformylase-like protein-like; polypeptide deformylase; similar to peptide deformylase-like protein | |
Mouse | |
Affinity Chromatography | |
RUO | |
64146 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |