Learn More
Invitrogen™ PDGF-BB Canine ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ ECPDGFB
Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.
Description
The Canine Platelet Derived Growth Factor BB (Dog PDGF-BB) ELISA quantitates Dog PDGF-BB in canine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Dog PDGF-BB. Principle of the method The Canine PDGF-BB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer or as a heterodimer with the platelet-derived growth factor alpha polypeptide , where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
Q6Q7I7 | |
4 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
442986 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Extracellular Matrix & Cell Adhesion | |
2°C to 8°C | |
Canine | |
4 hr. 45 min. |
4.92-1200 pg/mL | |
4pg/mL to 1,200pg/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA pair antibody detects canine PDGF-BB, it also can recognize human PDGF-BB. | |
Canine | |
Colorimetric Microplate Reader | |
PDGF-B | |
<10% | |
HRP | |
RUO | |
Plasma, 5 μL; Serum, 5 μL; Supernatant, 100 μL | |
PDGF-B | |
1 hr. 20 min. |