missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDGF-BB Porcine ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ ESPDGFB

Code : NEW

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15471124

  • £530.00 / 1 set
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, diluents, wash buffer, chromogen, stop solution, and plate covers.

Description

Description

The Porcine Platelet-Derived Growth Factor BB (Porc PDGF BB) ELISA quantitates Porc PDGF BB in porcine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Porc PDGF BB. Principle of the method The Porcine PDGF BB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer or as a heterodimer with the platelet-derived growth factor alpha polypeptide , where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
Specifications

Specifications

P20034
10 pg/ml
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
100626465
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Cell Cycle & Proliferation
2°C to 8°C
Porcine
4 hr. 45 min.
10.24-2500 pg/ml
10pg/mL to 2,500pg/mL
Biotin
Growth Factors & Receptors
This ELISA pair antibody detects porcine and human PDGF-BB. Other species not determined.
Porcine
Colorimetric Microplate Reader
PDGF-2
<10%
HRP
RUO
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL
PDGFRbetaPGF-Rb
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers