missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human PDGFRB (PDGFR beta), His Tag Recombinant Protein
Human PDGFRB (PDGFR beta), His Tag Recombinant Protein
Brand: Thermo Scientific™ PV3591
Code : 50
Additional Details : Weight : 0.05250kg
Description
- PDGFRB is a receptor tyrosine kinase
- It has been implicated in atherogenesis and in cell transformation by the SIS oncogene
Specifications
NP_002600 | |
66.3 kDa | |
PDGFRB (PDGFR β) | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic | |
Kinases & Inhibitors |
Kinase Assay | |
1 mg | |
PDGFR Kinase Family | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |