missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human PDGFRB (PDGFR beta), His Tag Recombinant Protein
Human PDGFRB (PDGFR beta), His Tag Recombinant Protein
Brand: Thermo Scientific™ P3082
This item is not returnable.
View return policy
Description
PDGFRB is a receptor tyrosine kinase and has been implicated in atherogenesis and in cell transformation by the SIS oncogene.Specifications
NP_002600 | |
5159 | |
10 μg | |
PDGFR Kinase Family | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
PDGFRB | |
Baculovirus | |
Recombinant, Catalytic Domain, Cytoplasmic | |
Receptor Tyrosine Kinases |
Kinase Assay | |
66.3 kDa | |
PDGFRB (PDGFR β) | |
aa 558-1106 | |
Recombinant | |
Storage Buffer: 20mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% NP-40, 1mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
platelet-derived growth factor receptor, beta polypeptide | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction