missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemsplash PE Disposable Visitor Coat

Click to view available options
:
140 x 120 mm
Description
Visitors Polythene Coat
- 30 GSM Polyethylene Fabric
- Stud Fastening with Fold Over Collar
- Elasticated Cuffs
- Ideal for Non-Hazardous Environments
- Ideal for Food Industry
- 140 x 120 mm
- PFAS Free
Description: Visitors Polythene Coat with stud closure front, 20-micron weight. Suitable for use in a wide range of industries, with a foldaway pouch.
Specifications
Specifications
| Product Type | Light-duty Visitor Lab Coat |
| Size | Universal |
| Gender | Unisex |
| Material | Polyethylene, Non-Woven Polypropylene Laminate |
| Color | Blue |
| Closure Type | Press Studs |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Liquids |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction