missing translation for 'onlineSavingsMsg'
Learn More
SCAT™ Safety Canisters, HDPE-EC Product Code.: 10125663
In Stock
to see stock.
Add to basket

SCAT™ Safety Canisters, HDPE-EC

Product Code. 10125663
10 L, S60/61, Canister with Floater
Click to view available options
Capacity (Metric):
5 L
10 L
20 L
30 L
60 L
Closure Size:
S50
S60/61
S70/71
S90
Includes:
Canister
Canister with Blue Stripes on Grip, without UN-Y Approval
Canister with Floater
Canister with Green Stripes on Grip, without UN-Y Approval
Canister with Red Stripes on Grip, without UN-Y Approval
Canister with UN-Y Approval
Canister with Viewing Strip, UN-Y Approval
Canister with Yellow Stripes on Grip, without UN-Y Approval

Product Code. 10125663

Brand: SCAT™ 108042

In Stock
to see stock.
Add to basket
Add to basket

SCAT™ electrically conductive safety canisters are made of high quality HDPE material. Due to their grounding capability, flying sparks caused by electrical discharge can be prevented when disposing of flammable or explosive laboratory liquids.

  • Made of very robust and electrically conductive HDPE, which is very resistant to solvents commonly used in laboratories
  • Filling quantities from 10 L for use with HPLC systems up to massive 60 L drums for the collection of large quantities of containers (e.g. professional SymLine™ disposal systems)
  • Thread types S50, S60/61, S70/71 and S90 are commonly used in the laboratory and are all compatible
  • Safety Waste Caps range for the safe disposal of volatile, hazardous substances
  • Special safety features such as integrated visibility strips or floats ensure permanent fill level monitoring
  • Practical designs as space-saving canisters allow safe storage of liquid waste even in small areas or in hazardous goods cabinets

Specifications

Autoclavable Not Autoclavable
Capacity (English) 2.6 gal.
Capacity (Metric) 10 L
Certifications/Compliance UN-Y Approval
Closure Size S60/61
Color Black
Dimensions (L x W x H) 290 x 185 x 265 mm
For Use With (Application) Collection of Flammable Liquid Waste
For Use With (Equipment) SCAT™ Safety Funnels
Height (Metric) 265 mm
Includes Canister with Floater
Material HDPE
Product Type Canister
Width (Metric) 185 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SCAT™ Safety Canisters, HDPE-EC >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.