missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ PE IBC Station with Filling Area and PE Grid
The PE IBC Station is approved for the safe storage of water pollutants.
Brand: asecos™ 24160
Additional Details : Weight : 210.00000kg
Description
- Highly resistant against oil, acid, alkali, etc.
- Easy and safe in-house transport of the empty sump by forklift truck
Specifications
Blue | |
1000 L | |
IBC Spill Pallet | |
2460.00 mm | |
2460.00 mm | |
485.00 mm | |
1800.00 mm | |
210 kg |
264.2 gal. | |
Polyethylene | |
96.85 in. | |
96.85 in. | |
19.09 in. | |
70.87 in. | |
462.97 lb. |