missing translation for 'onlineSavingsMsg'
Learn More

Sartorius PE Seperator Gauze

PE Seperator Gauze is Agar media for colony counting by the membrane filtration method or via direct incubation.

Brand:  Sartorius 13420--58------K

Product Code. 15922480

  • £95.40 / Pack of 50
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Time- and cost-saving
  • Compliance with international standards and references
  • Consistent quality
  • The traditional culture media for microorganisms is agar media.
  • The media are available in two formats.
  • Agar media in tubes are for pouring agar plates.
  • The content of one tube is sufficient for two 90 mm or three 60 mm petri dishes.
  • Agar media in bottles are the cost-effective alternative for casting plates.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Disc
Non-sterile
Glass Microfiber Filters with Binder
58 mm
Glass Fiber
50 Pk.
2.3 in.
Certified Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers