Learn More
Fisherbrand™ PE Snap Cap, Transparent, for Snap Cap Vial
Polyethylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 22 08 0794
33.69 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Description
- PE Snap Cap
- Transparent
- Closed top
- Snap caps are easy to apply and easy to remove, eliminate the need for crimping or de-capping tools
Specifications
Transparent | |
Polyethylene | |
ND22 |
22 mm | |
Snap Cap |