missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ PE Sump Pallet without Grid
The PE Sump Pallet is approved for the safe storage of water pollutants.
Brand: asecos™ 40048
This item is not returnable.
View return policy
Description
- PE blue
- Grid: without
- Accessible by forklift
Specifications
Blue | |
220 litres | |
Painted Spill Sump | |
1330.00 mm | |
1330.00 mm | |
295.00 mm | |
1330.00 mm | |
36 kg |
58.1 gal. | |
Polyethylene | |
52.36 in. | |
52.36 in. | |
11.61 in. | |
52.36 in. | |
79.37 lb. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction