missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ PE Sump for Small Containers with PE-Grid
The PE Sump for Small Containers is approved for the safe storage of water pollutants.
Brand: asecos™ 40024
This item is not returnable.
View return policy
Description
- PE black
- Grid: PE
- Ideal for water pollutant storage.
Specifications
Black | |
30 L | |
Painted Spill Sump | |
592.00 mm | |
592.00 mm | |
180.00 mm | |
400.00 mm | |
4.5 kg |
7.9 gal. | |
Polyethylene | |
23.31 in. | |
23.31 in. | |
7.09 in. | |
15.75 in. | |
9.92 lb. |