Learn More
Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint
Especially for distillation and evaporation and provides for ease of sample removal. Lenz Laborglasinstrumente™ Pear-Shaped Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03031937
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10508631
  • £191.00 / Pack of 10
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12383
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
According to DIN 12383
Borosilicate Glass, Duran Glass
64 mm
100 mL
19/26 NS