Learn More
Cytiva PEEK Fingertip Union
Used as a connector for chromatography applications. Cytiva PEEK Fingertip Union 1/16 in. Male/M6 Female.
Brand: Cytiva 18-1112-58
Code : GX
Additional Details : Weight : 0.01080kg
Description
- 1/16 in. Male/M6 Female Complete
- Packsize 8 Material
- Polyetheretherketone (PEEK)
Specifications
1/16 in. Male to M6 Female |