missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PEEK Large RN Hub Needles

For applications that call for a non-metallic needle.

Brand:  Hamilton™ 8650-01

Product Code. 11550262

  • £247.00 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Plastic needles
  • Manufactured from PEEK HPLC tubing that is both chemically and physically robust
  • Sold in packs of 6

Specifications

Specifications

Autoclavable
0.8 mm
Non-metallic needle requirements
4/Pk.
Large RN Hubs
Gas sterilizable
1.6 mm
PEEK Needle
3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers