Learn More
Cytiva PEEK Male Connector
Connect tubing of different sizes. Cytiva Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
Brand:  Cytiva 18-1112-49
Code : GX
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 11370052
  • £207.00 / Each
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Description
Description
  • Chromatography accessory
  • Complete Packsize 2
  • Select from a variety of connections sizes and internal diamters 
  • For use with chromatography tubing
  • Connectors made of Polyetheretherketone (PEEK)
Specifications
Specifications
Male Connector
3/16 in. Male
PEEK
3/16 in. O.D. Tubing