missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical PEEK Tubing, 1/16 in. O.D., 10 ft
Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs
£33.20 - £70.59
Specifications
pH Range | 2 to 14 |
---|---|
Diameter (English) Outer | 1/16 in. |
Overall Length (English) | 10 ft. |
Material | PEEK |
Max. Operating Temperature (Metric) | 100°C |
Product Code | Brand | Color | Diameter (English) Inner | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (English) Inner | Price | Quantity & Availability | ||||
16652562
|
Trajan Scientific and Medical
1302002500-10F |
Natural | 0.0025 in. |
£48.05
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
15780999
|
Trajan Scientific and Medical
1302004006-10F |
Black | 0.004 in. |
£68.11
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
15721009
|
Trajan Scientific and Medical
1302005001-10F |
Red | 0.005 in. |
£70.59
Each |
In Stock
Log in to see stock. |
||||
15761009
|
Trajan Scientific and Medical
1302007002-10F |
Yellow | 0.007 in. |
£61.35
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
15701019
|
Trajan Scientific and Medical
1302010003-10F |
Blue | 0.010 in. |
£49.20
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
15741019
|
Trajan Scientific and Medical
1302013004-10F |
Orange | 0.013 in. |
£34.75
Each |
Estimated Shipment: 25-07-2024
Log in to see stock. |
||||
15781019
|
Trajan Scientific and Medical
1302015008-10F |
Gray | 0.015 in. |
£33.20
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
16692562
|
Trajan Scientific and Medical
1302015000-10F |
Natural | 0.015 in. |
£48.05
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
15721029
|
Trajan Scientific and Medical
1302020004-10F |
Orange | 0.020 in. |
£50.36
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
15701039
|
Trajan Scientific and Medical
1302030005-10F |
Green | 0.030 in. |
£50.36
Each |
In Stock
Log in to see stock. |
||||
15761029
|
Trajan Scientific and Medical
1302030001-10F |
Red | 0.030 in. |
£33.20
Each |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
15741039
|
Trajan Scientific and Medical
1302040000-10F |
Natural | 0.040 in. |
£46.13
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
Description
- Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
- Bio-inert ad bio-compatible
- Color coded to industry standards for easy identification of inner diameter
- Leach-free colorants
- Smooth inner walls and precise IDs
- Works well with either polymer fittings as well as stainless steel fittings
- Can be formed to specific customer needs
Specifications
2 to 14 | |
10 ft. | |
100°C |
1/16 in. | |
PEEK |
For research use only