missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical PEEK Tubing, 1/16 in. O.D., 5 ft

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

Brand:  Trajan Scientific and Medical 1302020004-5F

 View more versions of this product

Product Code. 15741029

  • £29.45 / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs
Specifications

Specifications

Orange
0.020 in.
5 ft.
6000 psi (414 bar)
2 to 14
1/16 in.
PEEK
100°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only