missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical PEEK Tubing, 1/32 in. O.D., 50 ft

Designed specifically for modern LC, LCMS and automation useage. A flexible alternative to stainless steel tubing in high pressure applications, it ensures reproducibility in your chromatography flow rates and results through smooth internal surface along with consistent and tight toleranced IDs

£294.00 - £312.00

Specifications

Diameter (English) Outer 1/32 in.
Overall Length (English) 50 ft.
Max. Operating Temperature (Metric) 100°C
View More Specs

Products 8
Product Code Brand Color Diameter (English) Inner Price Quantity & Availability  
Product Code Brand Color Diameter (English) Inner Price Quantity & Availability  
16642552
View Documents Promotion Details
Trajan Scientific and Medical
1301002500-50F
Natural 0.0025 in.
£294.00
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
16682552
View Documents Promotion Details
Trajan Scientific and Medical
1301004006-50F
Black 0.004 in.
£294.00
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
15790979
View Documents Promotion Details
Trajan Scientific and Medical
1301005001-50F
Red 0.005 in.
£312.00
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
15730989
View Documents Promotion Details
Trajan Scientific and Medical
1301007002-50F
Yellow 0.007 in.
£312.00
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
15770989
View Documents Promotion Details
Trajan Scientific and Medical
1301010003-50F
Blue 0.010 in.
£302.00
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
15710999
View Documents Promotion Details
Trajan Scientific and Medical
1301015008-50F
Gray 0.015 in.
£294.00
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
16622562
View Documents Promotion Details
Trajan Scientific and Medical
1301015000-50F
Natural 0.015 in.
£294.00
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
15750999
View Documents Promotion Details
Trajan Scientific and Medical
1301020004-50F
Orange 0.020 in.
£312.00
Each
Estimated Shipment: 27-11-2024
to see stock.
Add to basket
 
Description

Description

  • Excellent chemical resistance and inert to most commonly used solvents. Exceptions are very caustic reagents, strong acids and bases
  • Bio-inert ad bio-compatible
  • Color coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with either polymer fittings as well as stainless steel fittings
  • Can be formed to specific customer needs
Specifications

Specifications

1/32 in.
50 ft.
100°C
Videos
SDS
Documents

Documents

Product Certifications

For research use only