Learn More
Cytiva PEEK Union
Connect piping of varying sizes. Cytiva Life Sciences™ PEEK Unions are an important accessory for various scientific applications.
Brand: Cytiva 18385801
This item is not returnable.
View return policy
Description
- Packsize 5
- When ordered separately the connectors are black.
- Made of polyetheretherketone (PEEK)
Specifications
Union | |
M6 female to 1/16 in. Male | |
PEEK |