Learn More
Cytiva PEEK Union
Shop All Cytiva Products
Click to view available options
Connections:
1/16 Male
1/16 in. Female to 1/16 in. Female
5/16 in. Female to 1/16 in. Male
M6 female to 1/16 in. Male
Unit Size:
Pack of 5
Pack of 8
Description
- Packsize 5
- When ordered separately the connectors are black.
- Made of polyetheretherketone (PEEK)
Specifications
Specifications
| Type | Union |
| Connections | 1/16 Male |
| Material | PEEK |
| Diameter (Metric) Inner | 0.5 mm |