Learn More
Hanna Instruments™ Gel Filled PEI Body Combination pH Electrodes
Features a PEI body, double junction pH electrode with a BNC connector that has a design consideration that is ideal for field applications and general purpose use.
Brand:  Hanna Instruments™ HI1230B
Product Code. 10527912
  • £438.63 / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Is a PEI body, double junction pH electrode with a BNC connector. 
  • Electrode features a single ceramic junction in the outer reference cell 
  • Spherical pH sensing portion is made with general purpose glass and is protected by an extension of the durable PEI body
  • Design consideration is ideal for field applications as well as general purpose use
  • Uses general purpose (GP) glass, spherical bulb, polyetherimide (PEI) body, single ceramic frit, and double junction
Specifications
Specifications
BNC
120 mm
0°C to 80°C
12 mm
0 to 13
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ Gel Filled PEI Body Combination pH Electrodes > Length: 120mm