missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PELP1 Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

Brand:  Novus Biologicals™ NBP2-56265PEP

Product Code. 18223064

This item is not returnable. View return policy

Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PELP1. The PELP1 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications

Specifications

27043
>80% by SDS-PAGE and Coomassie blue staining
PBS and 1M Urea, pH 7.4.
PELP1
100 μl
E. coli
Human
Store at -20°C. Avoid freeze-thaw cycles.
Antibody Competition
27 kDa
RUO
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54214. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers