Learn More
Memmert™ Entry Port
Material: Silicone; For Use With: IPP30
Brand:  Memmert™ IPP30F0
Code : NEW
Additional Details : Weight : 1.00000kg
Product Code. 15312732
  • £104.98 / Each
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Description
Description
  • 23 mm clear diameter
  • For introducing connections at the side
  • Can be closed by flap
  • Position left centre/centre or centre top
Specifications
Specifications
IPP30
Silicone
Entry Port
23 mm