missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ CH-3 Listen Only Headset

PELTOR™ CH-3 Listen Only Headsets utilise the FLX2 cable system for a simple and reliable connection. They feature a speaker system that’s optimised for speech frequency in high noise environments and a modern cup design which provides high comfort and attenuation.

Brand:  3M™

1315.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.



Additional Details : Weight : 3.00000kg

 View more versions of this product

Product Code. 17889066

  • £1900.00 / Pack of 10
Estimated Shipment: 09-09-2024
to see stock.
Add to basket

Description

Description

  • Protect your hearing without losing the ability to hear conversation with PELTOR™ CH-3 Listen Only Headsets.
  • These hear-only headsets feature a CH-3 speaker system that’s optimized for the speech frequency in high noise environments and meets the efficiency demand from today's digital devices.
  • The FLX2 cable system provides a simple and reliable connection and the modern cup design offers high comfort and attenuation.
  • All critical parts are rigorously tested and easy to change without any special tooling which contributes to low maintenance costs.
  • Comes with a headband, neckband or helmet attachment.
  • Available in bright yellow for the standard model or bright blue to indicate Explosive atmosphere (Ex) approval.
Specifications

Specifications

Blue
Communication Headset
25 dB
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers