Learn More
3M™ PELTOR™ Earmuffs
PELTOR™ X2 Earmuffs are our innovative hearing protectors with a modern slim cup design.
Brand:  3M™ 7100123157
210.82 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17893246
Description
Description
  • For high performance hearing protection against moderate noise levels, use 3M™ PELTOR™ X2 Earmuffs.
  • 3M PELTOR Earmuffs X2 are colour-coded yellow, making it simple to select the correct level of protection from the X series range.
  • The headband model features an electrically insulated, twin headband design that helps reduce heat build-up on top of the head, and the stainless steel wire, coated with plastic makes the headband the most robust 3M has to offer.
  • It is both strong and comfortable as the flat wide headband has a soft plastic coating providing stable pressure on top of the head.
  • Carefully moulded sliders ensure the earmuffs are easy to adjust when on the head.
  • Wide, soft foam ear cushions help workers stay comfortable, while the slim cups offer improved compatibility with other safety equipment.
  • Offering a product to meet each users job or preference the helmet mounted version is available as P5 with the same dielectric performance as the headband model.
  • Alternatively, if the application does not require electrical insulation the helmet mounted P3 model and neckband is available.
  • The cushions and foam inserts, available as Hygiene Kits, are easy to replace if they become worn or damaged, helping extend the life of the product.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 dB
Oval
No
No
No
Uncorded
Reusable
Black
Neckband
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ PELTOR™ Earmuffs > Neckband