Learn More
3M™ PELTOR™ Hygiene Kit
3M™ PELTOR™ Hygiene Kit contains replacement sealing rings and muffler pads which can be replaced in a few easy steps.
Brand: 3M™ 7100101874
9.96 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Kit contains replacement sealing rings and muffler pads which can be replaced in a few easy steps
- Extends the life of earmuffs
- Ensures good function and hygiene
- Avoid wearing uncomfortable earmuffs
- Pack contains 2 x foam inserts and 2 sealing rings
Specifications
Hygiene Kit | |
Worktunes Headset | |
Foam Inserts and Sealing Rings |