missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ LiteCom Headset

3M™ PELTOR™ LiteCom Headsets are hearing protectors with a built-in pre-programmed two-way communication radio and an integrated noise cancelling microphone. They’re built for demanding environments to help you communicate efficiently over hazardous noise.

Brand:  3M™ 7000108542

492.63 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15769997

  • £647.00 / Each
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Hearing protector with built-in analogue two-way communication radio
  • 8 channels in the PMR 446 MHz band
  • 38 sub-channels
  • Noise cancelling waterproof microphone provides voice-operated transmission (VOX) for hands free use
  • Voice guided menu system
  • Auto power off function
  • Clear battery status indication
  • Communication radio in the PMR 446 MHz frequency range.
  • Use to help you communicate efficiently in high noise environments

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Communication Headset
Neckband
32 dB
CE Approved
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ PELTOR™ LiteCom Headset > Model: Neckband

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.