missing translation for 'onlineSavingsMsg'
Learn More
3M™ PELTOR™ ProTac™ III Headset Product Code.: 15648175
Estimated Shipment: 03-06-2025
Add to basket

3M™ PELTOR™ ProTac™ III Headset

Product Code. 15648175
PELTOR™ ProTac III Headset, Hard Hat, 23 dB
Click to view available options
Model:
PELTOR™ ProTac III Headset
PELTOR™ ProTac III Slim Headset
Style:
Hard Hat
Headband
Noise Reduction Rating (NRR):
19 dB, 21 dB
23 dB
26 dB
This item is not returnable. View return policy

Product Code. 15648175

Brand: 3M™ 7100088423

Estimated Shipment: 03-06-2025
Add to basket
Add to basket
This item is not returnable. View return policy

Headset with level-dependent function for ambient listening helps you focus on your work and communicate with colleagues in noisy environments, without removing the hearing protection. Stereo feedback allows one to better determine direction of alarms, warning cries, backing vehicles and so on.

  • Hearing protector helps protect against harmful noise, whilst allowing ambient sounds to be heard
  • Built-in level-dependent function allows the wearer to hear ambient sounds, such as conversations, machine sounds and warning signals
  • 3.5 mm stereo jack to connect to your mobile phone or two-way radio to hear the call right in your ear
  • Auto power off turns headset off after 4 hours of non-use, to save energy and a low-battery warning informs you when to change batteries
  • Voice guided menu system so no need to take your headset off
TRUSTED_SUSTAINABILITY

Specifications

Color Black, White
Hearing Protection Style Earmuffs
For Use With (Application) Agriculture
Model PELTOR™ ProTac III Headset
Noise Reduction Rating (NRR) 23 dB
Product Type Protective Hearing Communications Headset
Style Hard Hat
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ PELTOR™ ProTac™ III Headset >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.