Learn More
3M™ PELTOR™ Rechargeable Li-ion Battery Pack
3M™ PELTOR™ Rechargeable Li-ion Battery Pack designed for use with 3M™ PELTOR™ LiteCom Plus Headsets.
Brand: 3M™ 7100075380
93.42 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Rechargeable Lithium-Ion battery pack
- Battery Capacity 3.7 V, 1350mAh, 5 Wh
- For use with 3M™ PELTOR™ LiteCom Plus headsets
- Charging time 4 hours
Specifications
Battery Pack |