missing translation for 'onlineSavingsMsg'
Learn More

3M™ PELTOR™ SportTac™ Replacement Shells

Product Code. p-7160324
Click to view available options
Color:
Black
Red
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15709937

Brand: 3M™ 7000107843

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

3M™ PELTOR™ SportTac™ Replacement Shells are interchangeable shells that makes it easy to change the color of earmuffs.

  • Clip on replacement shells for 3M™ PELTOR™ SportTac™ Hearing Protectors
  • Interchangeable shells makes it easy to change color
  • One pair per pack
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Product Type Shells
For Use With (Equipment) PELTOR™ SportTac™ Hearing Protectors
Includes One Pair
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.