missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ Sporttac TIR Sound Modulation Ear Muff
Klicka för att se tillgängliga alternativ
Color:
Black, Red
Förpackningsstorlek:
Each
Specifikationer
Specifikationer
| Color | Black, Red |
| For Use With (Application) | Designed for Hunters and Shooting Sports Enthusiasts |
| Plug Shape | Oval |
| Product Type | Tactical Sporttac TIR Sound Modulator |
| Includes | Integrated Automatic Shutdown Function |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction