Learn More
3M™ PELTOR™ Wireless Communication Accessory
Click to view available options
Quantity: 10
Description
- Communicate safely in noisy environments
- Features a noise cancelling microphone
- Bluetooth™ wireless technology for connecting to your smart phone
- Resistant to harsh environmental elements including temperature fluctuations, sweat, and rain
- Designed to be comfortable for all day wearability
- Easily attaches to 3M™ PELTOR™ X Series Earmuffs transforming them into a communication device
Specifications
Specifications
Color | Black |
Certifications/Compliance | IP54, eClass 9: 40019116 |
Product Type | Accessory |