missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ WS™ LiteCom PRO III Ex Headset
3M™ PELTOR™ WS™ LiteCom PRO III Ex Headset offers wireless communication in noisy environments.
Brand: 3M™ 7100098898
1330.25 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Built-in programmable two-way communication radio allows for clear, hands-free communication in high levels of noise without cumbersome cables or cords
- Headset can be programmed to use up to 70 unique two-way radio channels using analog and/or DMR digital modulation
- Bluetooth™ multipoint functionality
- Noise attenuating ear cups help provide hearing protection in environments with potentially hazardous noise
- Approved for use in potentially explosive atmospheres in accordance with the ATEX directive and the IECEx scheme for dust, gas and underground mining
- Level-dependent function for ambient listening with Push-to-Listen functionality
- Noise-cancelling waterproof speech microphone with voice operated transmission (VOX) for hands-free operation; helps provide clear communication in noisy and demanding environments

Specifications
Neckband | |
MT73H7B4D10EU-50 | |
Analog, Digital | |
32 dB |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3M™ PELTOR™ WS™ LiteCom PRO III Ex Headset > Model: MT73H7B4D10EU-50
Spot an opportunity for improvement?Share a Content Correction