Learn More
3M™ PELTOR™ WS™ ProTac XP Bluetooth™ Headset
3M™ PELTOR™ WS™ ProTac XP Bluetooth™ Headset for cable free clear and reliable communications with phones or other communication devices.
£443.65 - £497.00
Specifications
Product Type | Bluetooth™ Headset |
---|---|
Certifications/Compliance | CE Approved |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
15740018
|
3M™
7000147315 |
MT15H7P3EWS5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15750018
|
3M™
7000147316 |
MT15H7AWS5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15760018
|
3M™
7000147318 |
MT15H7BWS5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Active-hearing headset with Bluetooth™, for cable free clear and reliable communications with phones or other communication devices
- Level dependent function lets the user hear surrounding noise and converse with colleagues
- Work with both hands free while taking calls via DECT- or mobile phones
- Headset provides effective noise attenuation when sound level is high
- Supports Bluetooth™ A2DP profile for streaming high quality stereo sound
- VOX function for hands-free operation of the communication radio
Specifications
Bluetooth™ Headset | |
CE Approved |