missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Pendulum Beaker PP for Buerkle™ Telescopic Rod

This beaker automatically swings to a horizontal position. Nothing can be spilt. Suitable therefore for deep and narrow shafts (150 mm dia.). Designed for use with the Buerkle™ Telescopic Rod.

Brand:  Buerkle™ 5354-5100

49.31 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.



Code : 04

Additional Details : Weight : 0.10980kg

 View more versions of this product

Product Code. 10222371

  • £90.95 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • Polypropylene
  • For industry and water treatment
Specifications

Specifications

Polypropylene
1000 mL
Pendulum
For Industry and Water Treatment
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Exchangeable tool for Buerkle™ Telescopic Rod.