missing translation for 'onlineSavingsMsg'
Learn More

Penicillin G potassium salt, 99.9%, MP Biomedicals™

Product Code. 11477600
Click to view available options
Quantity:
10 mU
100 mU
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 113-98-4
Molecular Formula C33H36ClKN4O4S2
Molecular Weight (g/mol) 691.34
MDL Number MFCD00036193
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
Synonym Benzylpenicillin potassium salt
PubChem CID 23664709
ChEBI CHEBI:7963
IUPAC Name potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Penicillin G Potassium Salt
Physical Form Powder
Assay Percent Range 0.999
Formula Weight 372.5
Color White
Quantity 10 mU
Chemical Name or Material Penicillin G potassium salt, Potency: 1500-1700 u/mg
compliance-icons compliance-icons
Product Identifier
  • Penicillin G potassium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P321-Specific treatment (see on this label).
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Penicillin G potassium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'